Order Entry
Export
Orders LinkContactUsLinkComponent
SEAP assay kit

SEAP assay kit

SEAP assay kit, 1 KIT
Catalog # OZBISP00500
Supplier:   OZ BIOSCIENCES
SEAP assay kit
Catalog # OZBISP00500
Supplier:  OZ BIOSCIENCES
CAS Number:  
SEAP assay kit
SEAP assay kit

SEAP assay kit, 1 KIT

CAS Number:  
Price
151,73 €
Log in for availability
1 * 1 KIT
Product Details & Documents

SEAP assay kit (Secreted Alkaline Phosphatase) is a colorimetric assay for sensitive quantification of SEAP in culture medium from transfected cells or tissues.

  • Designed to detect SEAP reporter gene expression
  • Fast and easy analysis
  • Colorimetric-measure with a standard spectrophotometer or an ELISA reader (405 nm)
  • Multiple samples can be analysed in low volumes
  • Convenient and economical packaging: no tablet, no powder

Alkaline phosphatase cleaves the colourless 4-nitrophenylphosphate (pNPP) substrate to give a yellow p-nitrophenolate product in alkaline solutions. It is commonly used as a reporter gene expression and can be accurately quantified. It is a truncated form of placental alkaline phosphatase (GPI-anchored protein).

Delivery information: Kit contains pNPP-containing assay buffer component A (10×1 ml), reaction buffer component B (125 ml), reaction buffer component C (125 µl) and dilution buffer component D (50 ml).

Caution: Store at +4 °C.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    SEAP assay kit

Specifications