Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Thiazole orange homodimer (TOTO®-1)
Thiazole orange homodimer (TOTO®-1) 1 mM in DMSO
Thiazole orange homodimer (TOTO®-1) 1 mM in DMSO
Thiazole orange homodimer (TOTO®-1) 1 mM in DMSO
Catalog # 40079.
Supplier:
Biotium
CAS Number:
Expand image
Thiazole orange homodimer (TOTO®-1) 1 mM in DMSO
Catalog # 40079.
Supplier:
Biotium
CAS Number:
Specifications
Pk:
200 µl
Seq:
1358
Specifications
About this item
Product Details
:
:
1303 g/mol
CAS Number:
143413-84-7
Formula:
C₄₉H₅₈I₄N₆S₂
Product Details