ESD safety shoes, lace-up, Wellmaxx, Miles, Rob
Supplier: ELTEN
Certificates
About this item
Lightweight and comfortable ESD safety shoes made from cowhide leather/hydrophobized nubuck leather, full padded bellows tongue plus collar and a dual-density TPU/PU sole WELLMAXX.
- Climate regulating, breathable textile lining
- Replaceable insole; orthopaedic insoles possible
- Full-length inlay sole ESD PRO black
- Soft fleece ESD insole
- Scuff cap for additional protection
- Cushioning midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving the energy back to the wearer
- Puncture resistant steel midsole
- Protective composite toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/grey
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC
Specifications
- Catalog No:
- ELTN729451/42
- ELTN729451/43
- ELTN729451/40
- ELTN729451/41
- ELTN729451/45
- ELTN729451/46
- ELTN729451/47
- ELTN729451/48
- ELTN729451/44
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001