Order Entry
Export
ContactUsLinkComponent
3,4-Dichloroisocoumarin 98%
Toxic
Irritant
3,4-Dichloroisocoumarin 98%
Catalog # J65198.MA
CAS Number:  
Toxic
Irritant
3,4-Dichloroisocoumarin 98%
Catalog # J65198.MA
CAS Number:  

Specifications

  • Pk:
    10 mg

Specifications

About this item

 :
 :215.029 g/mol
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
 :GHS06
 :GHS07
CAS Number:51050-59-0
Formula:C₉H₄Cl₂O₂
MDL Number:MFCD00036960