Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Water
Water nuclease-free
Water nuclease-free
NEWEB1500S
:
NEW ENGLAND BIOLABS
:
Water nuclease-free
NEWEB1500S
:
NEW ENGLAND BIOLABS
:
Pk:
25 mL
Seq:
2315
:
:
18.02 g/mol
:
Freezer
:
7732-18-5
:
H₂O
:
MFCD00011332