0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Magnesium XL FS
Magnesium XL FS
Corrosive
Magnesium XL FS
Catalog # DIAS146109990314
Supplier:
DIAGNOSTIC SYSTEMS DIASYS
CAS Number:
Expand image
Corrosive
Magnesium XL FS
Catalog # DIAS146109990314
Supplier:
DIAGNOSTIC SYSTEMS DIASYS
CAS Number:
Specifications
Pack type:
Vial
Pk:
12 Vial
Seq:
0273
Pk Info:
12 x 25 ml
Specifications
About this item
Product Details
:
:
GHS05
Product Details