Order Entry
Export
ContactUsLinkComponent
Hand disinfectant balm, Baktolan® Balm
Hand disinfectant balm, Baktolan® Balm
Catalog # BODL9801440
Supplier:  Bode
Hand disinfectant balm, Baktolan® Balm
Catalog # BODL9801440
Supplier:  Bode

Specifications

  • Description:
    Hand disinfectant balm, Baktolan® balm, 350 ml
  • Pk:
    350 mL

Specifications

About this item

Baktolan® Balm is a water-in-oil balm with high quality nourishing components that were particularly adapted to the skin’s pH value (5,5). This stabilises the skin’s protective acid mantle. The ingredients vitamin E, allantoin and panthenol moisturise the skin intensively, providing a smoothing effect, support the cell regeneration and relieve skin irritation and redness of the skin.

  • Vitamin E, allantoin and panthenol moisturise the skin intensively
  • Increases skin firmness and keeps skin smooth
  • Promotes skin regeneration
  • Preserves the protective acid mantle of the skin
  • Relieve of skin irritation and redness
  • Pleasant scent, free of parabens

Ideal for sensitive skin. A repeated skin care with Baktolan balm during the day can reduce the risk of skin irritations significantly.

Caution: Use biocides safely. Always read the label and product information before use.

*The product(s) on this page are outside the scope of the EU Directive 2019/904