ESD safety shoes, lace-up, Wellmax, Reaction XXT
Supplier: ELTEN
Certificates
About this item
Metal-free ESD safety shoes made from waxy cowhide leather, full, padded bellows tongue plus a dual-density PU/PU sole (Wellmaxx Trainers).
- Climate-regulating, breathable textile lining
- Replaceable insole; orthopaedic insoles possible
- Full length inlay sole ESD PRO black
- Anatomically-shaped insole and ankle support for improved stability
- Cushioned midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving energy back to the wearer
- Abrasion-proof scuff cap
- Metal-free, puncture resistant midsole
- Composite toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC
Specifications
- Catalog No:
- ELTN729701/41
- ELTN729701/40
- ELTN729701/39
- ELTN729701/38
- ELTN729701/37
- ELTN729701/48
- ELTN729701/36
- ELTN729701/47
- ELTN729701/46
- ELTN729701/45
- ELTN729701/44
- ELTN729701/43
- ELTN729701/42
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001