Order Entry
Export
ContactUsLinkComponent
Anti-LAMA1, LAMB1, LAMC1 Rabbit Polyclonal Antibody
Anti-LAMA1, LAMB1, LAMC1 Rabbit Polyclonal Antibody
Catalog # BSENR-1808-100
Supplier:  Biosensis
Anti-LAMA1, LAMB1, LAMC1 Rabbit Polyclonal Antibody
Catalog # BSENR-1808-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Laminin-111
  • Antigen symbol:
    LAMA1, LAMB1, LAMC1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Format:
    Purified IgG
  • Form:
    Lyophilized
  • UniProtKB:
    Uniprot # P19137; Uniprot # P02469; Uniprot # P02468
  • Storage temperature:
    Store lyophilised antibody at 2-8 °C. After reconstitution divide into aliquots and store at –20 °C for long-term storage. Store at 2-8 °C short-term (up to 4 weeks) with an appropriate antibacterial agent. Avoid repetitive freeze/thaw cycles.
  • Shipping temperature:
    Ambient
  • Immunogen:
    Laminin-111 isolated from mouse EHS cells
  • Purification:
    Affinity purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Binding to cells via a high affinity receptor, laminin is thought to mediate the attachment, migration and organisation of cells into tissues during embryonic development by interacting with other extracellular matrix components. Ref: uniprot.org. This antibody recognizes laminin isotypes alpha-1 (440 kDa), beta-1 (220 kDa) and gamma-1 (220 kDa). It also binds laminin binding protein at 120 kDa which always co-expresses with laminin.

Western blotting (1:1,000-1:5,000) and Immunohistochemistry (1:1,000-1:5,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: LAMA1, LAMB1, LAMC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat