

tert-Butyl glycidyl ether ≥99%, Sigma-Aldrich®
Catalog # SIAL251712-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Assay:99%
- IR Spectrum:Conforms to structure
Specification Test Results
About this item
:
:130.19 g/mol
Density:0.917 g/ml (25 °C) (lit.)
EINECS:231-640-0
ADR:3,III
:M-Clarity™ quality level = MQ100
Flash Pt:41 °C
UN:3271
:GHS02
:GHS07
CAS Number:7665-72-7
Formula:C₇H₁₄O₂
MDL Number:MFCD00010050