Orders LinkContactUsLinkComponent
Safety ankle boots, lace-up, Basic Mid, Jori
   Sustainable Options Available
Supplier:   ELTEN
Safety ankle boots, lace-up, Basic Mid, Jori
Supplier:  ELTEN
   Sustainable Options Available
Certificates
Safety ankle boots, lace-up, Basic Mid, Jori
7 Options Available Below
Product Details & Documents
Extremely lightweight safety ankle boot made from cowhide leather, reflective materials, skin friendly textile lining, a full padded bellows tongue plus collar padding and a dual-density PU/PU jo_Basic sole (oil, fuel and abrasion resistant, also heat resistant up to approx. 130 °C).
  • Climate-regulating, breathable textile lining
  • Full length inlay sole
  • PU scuff cap protects the upper material
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
 : EN ISO 20345; S3, SRC
Specifications for Product Family
Specifications
  • Catalog No:
  • ELTN16331/44
  • ELTN16331/47
  • ELTN16331/48
  • ELTN16331/36
  • ELTN16331/37
  • ELTN16331/38
  • ELTN16331/40
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001
  • ISO 14001
  • ISO 14001
  • ISO 14001
  • ISO 14001
  • ISO 14001
  • ISO 14001
Specifications
Product Family Options
Product Information
  • Shoe size
    Colour
    Availability
    Price
  • 44
    Black
    Log in for availability
    Log in for availability
    1 * 1 PAIR
    1 * 1 PAIR - 32,90 €
    32,90 €
    1 * 1 PAIR
       32,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ELTN16331/44
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001
      Low Manufacturing Impact
  • 47
    Black
    Log in for availability
    Log in for availability
    1 * 1 PAIR
    1 * 1 PAIR - 32,90 €
    32,90 €
    1 * 1 PAIR
       32,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ELTN16331/47
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001
      Low Manufacturing Impact
  • 48
    Black
    Log in for availability
    Log in for availability
    1 * 1 PAIR
    1 * 1 PAIR - 32,90 €
    32,90 €
    1 * 1 PAIR
       32,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ELTN16331/48
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001
      Low Manufacturing Impact
  • 36
    Black
    Log in for availability
    Log in for availability
    1 * 1 PAIR
    1 * 1 PAIR - 32,90 €
    32,90 €
    1 * 1 PAIR
       32,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ELTN16331/36
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001
      Low Manufacturing Impact
  • 37
    Black
    Log in for availability
    Log in for availability
    1 * 1 PAIR
    1 * 1 PAIR - 32,90 €
    32,90 €
    1 * 1 PAIR
       32,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ELTN16331/37
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001
      Low Manufacturing Impact
  • 38
    Black
    Log in for availability
    Log in for availability
    1 * 1 PAIR
    1 * 1 PAIR - 32,90 €
    32,90 €
    1 * 1 PAIR
       32,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ELTN16331/38
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001
      Low Manufacturing Impact
  • 40
    Black
    Log in for availability
    Log in for availability
    1 * 1 PAIR
    1 * 1 PAIR - 32,90 €
    32,90 €
    1 * 1 PAIR
       32,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ELTN16331/40
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001
      Low Manufacturing Impact