Winter safety ankle boots, lace-up/Zip, Basic, Jerry, Jori
Supplier: ELTEN
Certificates
About this item
Warm winter safety boot feature a zip/lace-up combination that are made from cowhide leather, warm winter lining, a full padded bellows tongue, collar padding plus a dual-density PU/PU jo_Basic sole (oil, fuel and abrasion resistant, also heat resistant up to approx. 130 °C).
- Warm, moisture-absorbent lining
- Full length inlay sole
- Anti-static soft fleece insole
- PU scuff cap protects the upper material
- Puncture resistant steel midsole
- Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifications: EN ISO 20345; S3, SRC
Specifications
- Catalog No:
- ELTN16011/40
- ELTN16011/39
- ELTN16011/44
- ELTN16011/36
- ELTN16011/43
- ELTN16011/42
- ELTN16011/41
- ELTN16011/38
- ELTN16011/48
- ELTN16011/47
- ELTN16011/46
- ELTN16011/37
- ELTN16011/45
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001