Anti-CD2 Rabbit Monoclonal Antibody [clone: EP222]
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: EP222
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: EP222
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human