Anti-SLC45A3 Rabbit Monoclonal Antibody [clone: ZR9]
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: SLC45A3
Clonality: Monoclonal
Clone: ZR9
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human