Hydrotalcite (synthetic), Sigma-Aldrich®
SIAL652288-1KG
: Merck
:
- Pack type:Poly bottle
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
- Appearance (Color):White
- Appearance (Form):Powder
- X-Ray Diffraction:Conforms to Structure
- Ratio MgO/Al2O3:4.0 - 5.0
- Loss on Drying:< 0.35 %
: Dialuminum hexamagnesium carbonate hexadecahydroxide hydrate, Magnesium aluminum hydroxide carbonate hydrate, Magnesium aluminum hydroxycarbonate hydrate
:
:603.973 g/mol (anhy)
:Ambient
:11097-59-9
:12304-65-3
:CH₁₆Al₂Mg₆O₁₉.4H₂O
:MFCD01746913
:2.0 g/mL (25 °C) (lit.)
:M-Clarity™ quality level = MQ100