2-Adamantanol, Sigma-Aldrich®
SIAL153826-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
:
:152.24 g/mol
:299…303 °C
:1.07 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ100
:Ambient
:211-846-7
:700-57-2
:C₁₀H₁₆O
:MFCD00074744