Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Dibromothiophene, Sigma-Aldrich®

2,3-Dibromothiophene, Sigma-Aldrich®

Glass bottle for solids, 5 g
Catalog # SIALD43905-5G
Supplier:   Merck
2,3-Dibromothiophene, Sigma-Aldrich®
Catalog # SIALD43905-5G
Supplier:  Merck
CAS Number:  3140-93-0
2,3-Dibromothiophene, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS Number:  3140-93-0
Price
94,80 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:3140-93-0
  • Formula:C₄H₂Br₂S
  • MDL Number:MFCD00005418
  • Molecular Weight:241.93 g/mol
  • Boiling Pt:130 °C (1013 hPa)
  • Melting Pt:–17.5 °C
  • Density:2.2 g/cm³ (20 °C)
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ100
  •  :Density: 2.2 g/cm³ (20 °C)
  • EINECS:221-542-6
  • UN:1993
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications