

Hexamethylenetetramine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:100-97-0
:
Formula:C₆H₁₂N₄
MDL Number:MFCD00006895
Merck Index:13,05994
Melting Pt:280 °C
Density:1.33 g/cm³ (20 °C)
Flash Pt:250 °C
EINECS:202-905-8
UN:1328
ADR:4.1,III
:M-Clarity™ quality level = MQ200
:140.19 g/mol
:GHS02
:GHS07
Storage Temperature:Ambient