
2,4-Dichloro-5-methylpyrimidine, Sigma-Aldrich®
Catalog # SIALD66182-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:163.01 g/mol
Boiling Pt:235 °C (1013 hPa)
Melting Pt:26 °C
Density:1.39 g/cm³ (25 °C)
EINECS:217-227-8
UN:3261
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS05
CAS Number:1780-31-0
Formula:C₅H₄Cl₂N₂
MDL Number:MFCD00023197