Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Mercaptopyrimidine ≥98%

2-Mercaptopyrimidine ≥98%
Certificates
2-Mercaptopyrimidine ≥98%
2-Mercaptopyrimidine ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 1450-85-7 Molecular Formula: C4H4N2S Formula Weight: 112.15 Melting Point: ca 230° dec. Beilstein Reference: 107105

Synonyms:  2-Thiopyrimidine
  • CAS Number:1450-85-7
  • Formula:C₄H₄N₂S
  • MDL Number:MFCD00006073
  • Molecular Weight:112.16 g/mol
  • Melting Pt:∼ 230 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:215-917-3
Product Family Options

Product Information