
2,4,6-Triaminopyrimidine 97%
ACRO139250250
:
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
- Appearance:Almost white to yellow to light brown Powder
- Infrared spectrum:Conforms
- Melting point:246 to 252 °C
- Titration with HClO4:96.0 to 104.0 %
:
:125.13 g/mol
:324 °C (1013 hPa)
:246…252 °C
:1,45 g/cm³ (20 °C)
:Ambient
:213-720-7
:GHS07
:1004-38-2
:C₄H₇N₅
:MFCD00006100