
2,4-Dichloro-5-nitropyrimidine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:49845-33-2
:
Formula:C₄HCl₂N₃O₂
MDL Number:MFCD00127867
:193.98 g/mol
Boiling Pt:326 °C (1013 hPa)
Melting Pt:29.3 °C
Density:1.74 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:GHS07