Order Entry
Export
ContactUsLinkComponent
2,4-Dichloro-5-nitropyrimidine
Irritant
2,4-Dichloro-5-nitropyrimidine
Catalog # MOLEM65095899
Supplier:  Molekula
CAS Number:  
Irritant
2,4-Dichloro-5-nitropyrimidine
Catalog # MOLEM65095899
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :193.98 g/mol
Boiling Pt:326 °C (1013 hPa)
Boiling Pt:326 °C (1013 hPa)
Melting Pt:29.3 °C
Melting Pt:29,3 °C
Density:1,74 g/cm³ (20 °C)
 :GHS07
CAS Number:49845-33-2
Formula:C₄HCl₂N₃O₂
MDL Number:MFCD00127867