Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Dichloro-5-nitropyrimidine ≥97%

2,4-Dichloro-5-nitropyrimidine ≥97%

Glass bottle for solids, 10 g
Catalog # H27644.09
Irritant
2,4-Dichloro-5-nitropyrimidine ≥97%
Catalog # H27644.09
CAS Number:  49845-33-2
2,4-Dichloro-5-nitropyrimidine ≥97%
2,4-Dichloro-5-nitropyrimidine ≥97%

Glass bottle for solids, 10 g

CAS Number:  49845-33-2
Price
173,00 €
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 97% CAS No.: 49845-33-2 Molecular Formula: C4HCl2N3O2 Formula Weight: 193.98 Flash Point: 110°(230°F) Melting Point: 28-32° Boiling Point: 0.9°/0.5mm MDL No.: MFCD00127867

  • CAS Number:49845-33-2
  • Formula:C₄HCl₂N₃O₂
  • MDL Number:MFCD00127867
  • Molecular Weight:193.98 g/mol
  • Boiling Pt:0.9 °C (0.5 mmHg)
  • Melting Pt:28…32 °C
  • Density:1.74 g/cm³ (20 °C)
  • Flash Pt:>110 °C (230 °F)
  •  :Density: 1.74 g/cm³ (20 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications