Order Entry
Export
ContactUsLinkComponent
2-Bromopyrimidine
Irritant
2-Bromopyrimidine
Catalog # MOLE13695189-5G
Supplier:  Molekula
CAS Number:  
Irritant
2-Bromopyrimidine
Catalog # MOLE13695189-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Boiling Pt:62…64 °C (1.5 mmHg)
Boiling Pt:62…64 °C (1,5 mmHg)
Melting Pt:56…57.5 °C
Density:>1 g/cm³ (20 °C)
 :158.99 g/mol
Melting Pt:56…57,5 °C
Storage Temperature:Freezer
EINECS:224-993-7
 :GHS07
CAS Number:4595-60-2
Formula:C₄H₃BrN₂
MDL Number:MFCD00014601