
2,2'-Bipyridyl, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:366-18-7
:
Formula:C₁₀H₈N₂
MDL Number:MFCD00006212
:156.19 g/mol
Merck Index:13,03382
Boiling Pt:273 °C (1013 hPa)
Melting Pt:70…72 °C
Density:1.326 g/cm³ (–150 °C)
Storage Temperature:Ambient
EINECS:206-674-4
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:121 °C
:GHS06