2,2'-Bipyridyl
Glass bottle for solids, 100 g
Catalog # MOLEM30183563
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: α,α′-Dipyridyl, 2,2'-Bipyridine, 2,2'-Dipyridyl
- CAS Number:366-18-7
- Formula:C₁₀H₈N₂
- MDL Number:MFCD00006212
- Molecular Weight:156.19 g/mol
- Merck Index:13,03382
- Boiling Pt:273 °C (1013 hPa)
- Melting Pt:70…72 °C
- Density:1.326 g/cm³ (–150 °C)
- Flash Pt:121 °C
- Storage Temperature:Ambient
- EINECS:206-674-4
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



