2,2'-Bipyridyl ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99+% CAS No.: 366-18-7 Molecular Formula: C10H8N2 Formula Weight: 156.19 Flash Point: 121°(249°F) Melting Point: 70° Boiling Point: 272-273° Beilstein Reference: 113089 Merck Reference: 14,3347 UN No.: 2811 MDL No.: MFCD00006212
Synonyms: α,α′-Dipyridyl, 2,2'-Bipyridine, 2,2'-Dipyridyl
- CAS Number:366-18-7
- Formula:C₁₀H₈N₂
- MDL Number:MFCD00006212
- Molecular Weight:156.19 g/mol
- Merck Index:14,3347
- Boiling Pt:272…273 °C
- Melting Pt:70 °C
- Density:1.326 g/cm³ (–150 °C)
- Flash Pt:121 °C (249 °F)
- Storage Temperature:Ambient
- EINECS:206-674-4
- UN:2811
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Purity: 99+% CAS No.: 366-18-7 Molecular Formula: C10H8N2 Formula Weight: 156.19 Flash Point: 121°(249°F) Melting Point: 70° Boiling Point: 272-273° Beilstein Reference: 113089 Merck Reference: 14,3347 UN No.: 2811 MDL No.: MFCD00006212
Synonyms: α,α′-Dipyridyl, 2,2'-Bipyridine, 2,2'-Dipyridyl
- CAS Number:366-18-7
- Formula:C₁₀H₈N₂
- MDL Number:MFCD00006212
- Molecular Weight:156.19 g/mol
- Merck Index:14,3347
- Boiling Pt:272…273 °C
- Melting Pt:70 °C
- Density:1.326 g/cm³ (–150 °C)
- Flash Pt:121 °C (249 °F)
- Storage Temperature:Ambient
- EINECS:206-674-4
- UN:2811
- ADR:6.1,III



