2,6-Lutidine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-48-5
:
Formula:C₇H₉N
MDL Number:MFCD00006345
:107.16 g/mol
Merck Index:13,05636
Boiling Pt:143.7 °C (1013 hPa)
Melting Pt:–6 °C
Density:0.9252 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-587-3
UN:1993
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:33 °C