2,3-Lutidine, Sigma-Aldrich®
Catalog # SIALL3501-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:107.16 g/mol
Boiling Pt:161 °C (1013 hPa)
Melting Pt:–15 °C
Density:0.945 g/cm³ (20 °C)
EINECS:209-514-1
UN:2929
ADR:6.1,II
:M-Clarity™ quality level = MQ100
CAS Number:583-61-9
Formula:C₇H₉N
MDL Number:MFCD00009605