Order Entry
Export
ContactUsLinkComponent
2,4-Dihydroxypyridine, Sigma-Aldrich®
Irritant
2,4-Dihydroxypyridine, Sigma-Aldrich®
Catalog # SIAL176974-5G
Supplier:  Merck
CAS Number:  
Irritant
2,4-Dihydroxypyridine, Sigma-Aldrich®
Catalog # SIAL176974-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :111.10 g/mol
Boiling Pt:315 °C (1013 hPa)
Melting Pt:275…277 °C
 :M-Clarity™ quality level = MQ200
EINECS:210-924-8
 :GHS07
CAS Number:626-03-9
 :84719-31-3
Formula:C₅H₅NO₂
MDL Number:MFCD00006273