2,4-Dihydroxypyridine 97%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
2,4-Dihydroxypyridine 97%
Catalog # ACRO175590050
Supplier: Thermo Fisher Scientific
CAS Number: 626-03-9
Jump to:
Product Details & Documents
Synonyms: Pyridine-2,4-diol, 2,4-Pyridindiol, 3-Deazauracil, 4-Hydroxy-2-pyridone
- CAS Number:626-03-9
- Formula:C₅H₅NO₂
- MDL Number:MFCD00006273
- Molecular Weight:111.10 g/mol
- Boiling Pt:315 °C (1013 hPa)
- Melting Pt:278 °C (dec.)
- Storage Temperature:Ambient
- EINECS:210-924-8
Documents
ACRO17559
SDS
Specifications
Specification Test Results
- Appearance:White to yellow to light beige Crystalline powder or crystals
- Infrared spectrum:Conforms
- Titration with HClO4:≥96.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



