2-Hydroxypyridine
Glass bottle for solids, 100 g
Catalog # MOLEM32604337
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone
- CAS Number:142-08-5
- Formula:C₅H₅NO
- MDL Number:MFCD00006268
- Molecular Weight:95.10 g/mol
- Boiling Pt:324 °C (1013 hPa)
- Melting Pt:107 °C
- Density:1.085 g/cm³ (30 °C)
- Storage Temperature:Ambient
- EINECS:205-520-3
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



