Order Entry
Export
Orders LinkContactUsLinkComponent
Mouse Recombinant G-CSF (from <i>E. coli</i>)

Mouse Recombinant G-CSF (from E. coli)

Mouse Recombinant G-CSF (from <i>E. coli</i>)
Certificates
Mouse Recombinant G-CSF (from <i>E. coli</i>)

4 Options Available Below

Product Details & Documents

Granulocyte-colony stimulating factor (G-CSF) is a cytokine that functions as a potent inducer of neutrophilic granulocyte proliferation, terminal differentiation, and activation. G-CSF synthesis occurs in monocyte, macrophage, epithelial, endothelial, and fibroblast cells after activation by bacterial endotoxins, tumor necrosis factor alpha (TNF-α), interleukin 1 (IL-1), or interleukin 17 (IL-17). The functional activity of G-CSF is mediated through the granulocyte colony-stimulating factor receptor (G-CSF-R) to activate JAK/STAT and MAPK signal transduction pathways. G-CSF also promotes neurogenesis and inhibits neuronal apoptosis. Human and mouse G-CSF proteins are cross-reactive. 

  • High Quality
  • Low Endotoxin
  • Affordable

A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  

Caution: For research use only.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SHBT200-14-10UG
  • SHBT200-14-100UG
  • SHBT200-14-500UG
  • SHBT200-14-1MG
  • Protein/peptide name:
  • G-CSF
  • G-CSF
  • G-CSF
  • G-CSF
  • Protein synonyms:
  • GM-CSFβ|pluripoietin|MGI-1G|CSF-3
  • GM-CSFβ|pluripoietin|MGI-1G|CSF-3
  • GM-CSFβ|pluripoietin|MGI-1G|CSF-3
  • GM-CSFβ|pluripoietin|MGI-1G|CSF-3
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P09920
  • P09920
  • P09920
  • P09920
  • Sequence:
  • MVPLVTVSAL PPSLPLPRSF LLKSLEQVRK IQASGSVLLE QLCATYKLCH PEELVLLGHS LGIPKASLSG CSSQALQQTQ CLSQLHSGLC LYQGLLQALS GISPALAPTL DLLQLDVANF ATTIWQQMEN LGVAPTVQPT QSAMPAFTSA FQRRAGGVLA ISYLQGFLET ARLALHHLA
  • MVPLVTVSAL PPSLPLPRSF LLKSLEQVRK IQASGSVLLE QLCATYKLCH PEELVLLGHS LGIPKASLSG CSSQALQQTQ CLSQLHSGLC LYQGLLQALS GISPALAPTL DLLQLDVANF ATTIWQQMEN LGVAPTVQPT QSAMPAFTSA FQRRAGGVLA ISYLQGFLET ARLALHHLA
  • MVPLVTVSAL PPSLPLPRSF LLKSLEQVRK IQASGSVLLE QLCATYKLCH PEELVLLGHS LGIPKASLSG CSSQALQQTQ CLSQLHSGLC LYQGLLQALS GISPALAPTL DLLQLDVANF ATTIWQQMEN LGVAPTVQPT QSAMPAFTSA FQRRAGGVLA ISYLQGFLET ARLALHHLA
  • MVPLVTVSAL PPSLPLPRSF LLKSLEQVRK IQASGSVLLE QLCATYKLCH PEELVLLGHS LGIPKASLSG CSSQALQQTQ CLSQLHSGLC LYQGLLQALS GISPALAPTL DLLQLDVANF ATTIWQQMEN LGVAPTVQPT QSAMPAFTSA FQRRAGGVLA ISYLQGFLET ARLALHHLA
  • Protease-free:
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
  • Bioactivity assay: NFS-60 cell proliferation, ED50 ≤0,05 ng/ml, Bioactivity ≥2,0×10⁷ units/mg
  • Bioactivity assay: NFS-60 cell proliferation, ED50 ≤0,05 ng/ml, Bioactivity ≥2,0×10⁷ units/mg
  • Bioactivity assay: NFS-60 cell proliferation, ED50 ≤0,05 ng/ml, Bioactivity ≥2,0×10⁷ units/mg
  • Bioactivity assay: NFS-60 cell proliferation, ED50 ≤0,05 ng/ml, Bioactivity ≥2,0×10⁷ units/mg
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 10 mM sodium citrate, pH 3,0
  • 10 mM sodium citrate, pH 3,0
  • 10 mM sodium citrate, pH 3,0
  • 10 mM sodium citrate, pH 3,0
  • Molecular weight:
  • 19,1 kDa
  • 19,1 kDa
  • 19,1 kDa
  • 19,1 kDa
  • Reconstitution instructions:
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Storage conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • Tested applications:
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Vial
    10 µG
    Log in for availability
    Log in for availability
    1 * 10 µG
    1 * 10 µG - 190,00 €
    190,00 €
    1 * 10 µG
       190,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-14-10UG
    Protein/peptide name
    G-CSF
    Protein synonyms
    GM-CSFβ|pluripoietin|MGI-1G|CSF-3
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P09920
    Sequence
    MVPLVTVSAL PPSLPLPRSF LLKSLEQVRK IQASGSVLLE QLCATYKLCH PEELVLLGHS LGIPKASLSG CSSQALQQTQ CLSQLHSGLC LYQGLLQALS GISPALAPTL DLLQLDVANF ATTIWQQMEN LGVAPTVQPT QSAMPAFTSA FQRRAGGVLA ISYLQGFLET ARLALHHLA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: NFS-60 cell proliferation, ED50 ≤0,05 ng/ml, Bioactivity ≥2,0×10⁷ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium citrate, pH 3,0
    Molecular weight
    19,1 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG - 615,00 €
    615,00 €
    1 * 100 µG
       615,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-14-100UG
    Protein/peptide name
    G-CSF
    Protein synonyms
    GM-CSFβ|pluripoietin|MGI-1G|CSF-3
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P09920
    Sequence
    MVPLVTVSAL PPSLPLPRSF LLKSLEQVRK IQASGSVLLE QLCATYKLCH PEELVLLGHS LGIPKASLSG CSSQALQQTQ CLSQLHSGLC LYQGLLQALS GISPALAPTL DLLQLDVANF ATTIWQQMEN LGVAPTVQPT QSAMPAFTSA FQRRAGGVLA ISYLQGFLET ARLALHHLA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: NFS-60 cell proliferation, ED50 ≤0,05 ng/ml, Bioactivity ≥2,0×10⁷ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium citrate, pH 3,0
    Molecular weight
    19,1 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    500 µG
    Log in for availability
    Log in for availability
    1 * 500 µG
    1 * 500 µG - 2 660,00 €
    2 660,00 €
    1 * 500 µG
       2 660,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-14-500UG
    Protein/peptide name
    G-CSF
    Protein synonyms
    GM-CSFβ|pluripoietin|MGI-1G|CSF-3
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P09920
    Sequence
    MVPLVTVSAL PPSLPLPRSF LLKSLEQVRK IQASGSVLLE QLCATYKLCH PEELVLLGHS LGIPKASLSG CSSQALQQTQ CLSQLHSGLC LYQGLLQALS GISPALAPTL DLLQLDVANF ATTIWQQMEN LGVAPTVQPT QSAMPAFTSA FQRRAGGVLA ISYLQGFLET ARLALHHLA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: NFS-60 cell proliferation, ED50 ≤0,05 ng/ml, Bioactivity ≥2,0×10⁷ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium citrate, pH 3,0
    Molecular weight
    19,1 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 4 440,00 €
    4 440,00 €
    1 * 1 mg
       4 440,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-14-1MG
    Protein/peptide name
    G-CSF
    Protein synonyms
    GM-CSFβ|pluripoietin|MGI-1G|CSF-3
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P09920
    Sequence
    MVPLVTVSAL PPSLPLPRSF LLKSLEQVRK IQASGSVLLE QLCATYKLCH PEELVLLGHS LGIPKASLSG CSSQALQQTQ CLSQLHSGLC LYQGLLQALS GISPALAPTL DLLQLDVANF ATTIWQQMEN LGVAPTVQPT QSAMPAFTSA FQRRAGGVLA ISYLQGFLET ARLALHHLA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: NFS-60 cell proliferation, ED50 ≤0,05 ng/ml, Bioactivity ≥2,0×10⁷ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium citrate, pH 3,0
    Molecular weight
    19,1 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay