
Pyridine-2,3-dicarboxylic anhydride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:699-98-9
:
Formula:C₇H₃NO₃
MDL Number:MFCD00005915
:149.11 g/mol
Boiling Pt:360 °C (1013 hPa)
Melting Pt:146 °C
:M-Clarity™ quality level = MQ100
EINECS:211-834-1
:GHS07