Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pyridine-3,5-dicarboxylic acid
Pyridine-3,5-dicarboxylic acid ≥98%
Pyridine-3,5-dicarboxylic acid ≥98%
:
Thermo Fisher Scientific
Pyridine-3,5-dicarboxylic acid ≥98%
:
Thermo Fisher Scientific
:
499-81-0
:
:
C₇H₅NO₄
:
MFCD00006393
:
167.12 g/mol
:
357 °C (1013 hPa)
:
∼ 325 °C (dec.)
:
Ambient
:
207-893-8
:
GHS07
msds_ALFAA11540
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years