
Pyridine-3,4-dicarboxylic acid
Catalog # MOLE34910606-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:167.12 g/mol
Merck Index:13,02304
Boiling Pt:529 °C (1013 hPa)
Melting Pt:257 °C
Storage Temperature:Ambient
EINECS:207-705-4
:GHS07
CAS Number:490-11-9
Formula:C₇H₅NO₄
MDL Number:MFCD00006392