
Pyridine-3,4-dicarboxylic acid 97%
Catalog # ACRO131890250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to light beige Crystalline powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥96.0 %
- Titration with HClO4:≥97.5 %
Specification Test Results
About this item
:
:167.12 g/mol
Merck Index:13,02304
Boiling Pt:529 °C (1013 hPa)
Melting Pt:262 °C (dec.)
Storage Temperature:Ambient
EINECS:207-705-4
:GHS07
CAS Number:490-11-9
Formula:C₇H₅NO₄
MDL Number:MFCD00006392