
Pyridine-2,6-dicarboxylic acid
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:499-83-2
:
Formula:C₇H₅NO₄
MDL Number:MFCD00006299
:167.12 g/mol
Boiling Pt:357 °C (1013 hPa)
Melting Pt:327 °C
Flash Pt:188 °C
Storage Temperature:Ambient
EINECS:207-894-3
:GHS07