Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Aminonicotinic acid

Glass bottle for solids, 25 g
Catalog # MOLEM75348842
Supplier:   Molekula
2-Aminonicotinic acid
Catalog # MOLEM75348842
Supplier:  Molekula
CAS Number:  5345-47-1
2-Aminonicotinic acid

Glass bottle for solids, 25 g

CAS Number:  5345-47-1
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2-Aminopyridine-3-carboxylic acid, 2-Amino-3-pyridinecarboxylic acid
  • CAS Number:5345-47-1
  • Formula:C₆H₆N₂O₂
  • MDL Number:MFCD00006318
  • Molecular Weight:138.13 g/mol
  • Melting Pt:297 °C
  • Density:1.545 g/cm³ (20 °C)
  • EINECS:226-296-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications