
Methyl isonicotinate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2459-09-8
:
Formula:C₇H₇NO₂
MDL Number:MFCD00006427
:137.14 g/mol
Boiling Pt:207…209 °C
Melting Pt:8…8.5 °C
Density:1.016
Flash Pt:82 °C (180 °F)
EINECS:219-456-8
:GHS07
Storage Temperature:Refrigerator