Order Entry
Export
Orders LinkContactUsLinkComponent
 

6-Chloronicotinic acid

Irritant
Supplier:   Thermo Scientific
6-Chloronicotinic acid
Supplier:  Thermo Scientific
Certificates
6-Chloronicotinic acid

3 Options Available Below

Product Details & Documents
Synonyms:  6-Chloropyridine-3-carboxylic acid, 6-Chloro-3-pyridinecarboxylic acid
  • CAS Number:5326-23-8
  • Formula:C₆H₄ClNO₂
  • MDL Number:MFCD00006241
  • Molecular Weight:157.56 g/mol
  • Boiling Pt:284 °C (1013 hPa)
  • Melting Pt:188…189 °C
  • Storage Temperature:Ambient
  • EINECS:226-201-5
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g