Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Dichloro-4-methylpyridazine
3,6-Dichloro-4-methylpyridazine
3,6-Dichloro-4-methylpyridazine
Irritant
3,6-Dichloro-4-methylpyridazine
Supplier:
Thermo Scientific
Certificates
Expand image
Irritant
3,6-Dichloro-4-methylpyridazine
Supplier:
Thermo Scientific
About this item
CAS Number:
19064-64-3
:
Formula:
C₅H₄Cl₂N₂
MDL Number:
MFCD00006465
:
163.01 g/mol
Boiling Pt:
130…135 °C (1 mmHg)
Melting Pt:
87 °C
EINECS:
242-794-3
:
GHS07