
3,6-Dichloro-4-methylpyridazine ≥97%
Catalog # A19998.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:163.01 g/mol
Boiling Pt:149…151 °C (21 mmHg)
Melting Pt:86…88 °C
EINECS:242-794-3
:GHS07
Storage Temperature:Ambient
CAS Number:19064-64-3
Formula:C₅H₄Cl₂N₂
MDL Number:MFCD00006465