0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
3,6-Dichloropyridazine
3,6-Dichloropyridazine ≥98%
3,6-Dichloropyridazine ≥98%
:
Thermo Fisher Scientific
3,6-Dichloropyridazine ≥98%
:
Thermo Fisher Scientific
:
141-30-0
:
:
C₄H₂Cl₂N₂
:
MFCD00006466
:
148.98 g/mol
:
89…91 °C (0.2 mmHg)
:
66…70 °C
:
1.7 g/cm³ (20 °C)
:
Ambient
:
205-478-6
:
2811
:
6.1,III
:
GHS06
:
GHS07
msds_ALFAA14795
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years