
3,6-Dichloropyridazine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:141-30-0
:
Formula:C₄H₂Cl₂N₂
MDL Number:MFCD00006466
:148.98 g/mol
Boiling Pt:89…91 °C (0.2 mmHg)
Melting Pt:65…69 °C
Density:1,7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-478-6
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Off-white to brown Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:65 to 70 °C
- GC:≥96.0 %