
2,6-Dimethylpyrazine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-50-9
:
Formula:C₆H₈N₂
MDL Number:MFCD00006148
:108.14 g/mol
Boiling Pt:154 °C (1013 hPa)
Melting Pt:38…42 °C
Density:0.9647 g/cm³ (50 °C)
Flash Pt:52 °C
UN:1325
ADR:4.1,III
:M-Clarity™ quality level = MQ100
EINECS:203-589-4
:GHS07
Storage Temperature:Ambient