
2,5-Dimethylpyrazine 99%
:123-32-0
:
:C₆H₈N₂
:MFCD00006147
:108.14 g/mol
:155 °C (1013 hPa)
:15 °C
:0.99 g/cm³
:63 °C
:204-618-3
:GHS07
:Ambient
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4987 to 1.5007 (20°C, 589 nm)
- GC:≥98.5 %