2,3-Dimethylpyrazine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:5910-89-4
:
Formula:C₆H₈N₂
MDL Number:MFCD00006144
:108.14 g/mol
Boiling Pt:156 °C (1013 hPa)
Melting Pt:–12 °C
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:227-630-0
ADR:3,III
:M-Clarity™ quality level = MQ300
UN:1993