

2,3-Dimethylpyrazine ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5910-89-4
:
Formula:C₆H₈N₂
MDL Number:MFCD00006144
:108.14 g/mol
Boiling Pt:156 °C
Melting Pt:–12 °C
Density:1.022
Flash Pt:54 °C (129 °F)
Storage Temperature:Ambient
EINECS:227-630-0
UN:1993
ADR:3,III
:GHS02
:GHS07